MilliporeSigma
  • The White Collar protein WcoA of Fusarium fujikuroi is not essential for photocarotenogenesis, but is involved in the regulation of secondary metabolism and conidiation.
The White Collar protein WcoA of Fusarium fujikuroi is not essential for photocarotenogenesis, but is involved in the regulation of secondary metabolism and conidiation.
Fungal genetics and biology : FG & B (2008-01-22)
Alejandro F Estrada, Javier Avalos
ABSTRACT
The fungal proteins of the White Collar photoreceptor family, represented by WC-1 from Neurospora crassa, mediate the control by light of different biochemical and developmental processes, such as carotenogenesis or sporulation. Carotenoid biosynthesis is induced by light in the gibberellin-producing fungus Fusarium fujikuroi. In an attempt to identify the photoreceptor for this response, we cloned the only WC-1-like gene present in the available Fusarium genomes, that we called wcoA. The predicted WcoA polypeptide is highly similar to WC-1 and contains the relevant functional domains of this protein. In contrast to the Neurospora counterpart, wcoA expression is not affected by light. Unexpectedly, targeted wcoA disruptant strains maintain the light-induced carotenogenesis. Furthermore, the wcoA mutants show a drastic reduction of fusarin production in the light, and produce less gibberellins and more bikaverins than the parental strain under nitrogen-limiting conditions. The changes in the production of the different products indicate a key regulatory role for WcoA in secondary metabolism of this fungus. Additionally, the mutants are severely affected in conidiation rates under different culture conditions, indicating a more general regulatory role for this protein.
Sigma-Aldrich
Bikaverin, from Fusarium subglutinans, ≥98% (HPLC)