MilliporeSigma
  • Beneficial effect of spironolactone administration on ethynylestradiol-induced cholestasis in the rat: involvement of up-regulation of multidrug resistance-associated protein 2.
Beneficial effect of spironolactone administration on ethynylestradiol-induced cholestasis in the rat: involvement of up-regulation of multidrug resistance-associated protein 2.
Drug metabolism and disposition: the biological fate of chemicals (2007-08-10)
María L Ruiz, Silvina S M Villanueva, Marcelo G Luquita, Shin-ichi Ikushiro, Aldo D Mottino, Viviana A Catania
ABSTRACT
The effect of spironolactone (SL) administration on 17alpha-ethynylestradiol (EE)-induced cholestasis was studied, with emphasis on expression and activity of Mrps. Adult male Wistar rats were divided into the following groups: EE (5 mg/kg daily for 5 days, s.c.), SL (200 micromol/kg daily for 3 days, i.p.), EE+SL (same doses, SL administered the last 3 days of EE treatment), and controls. SL prevented the decrease in bile salt-independent fraction of bile flow induced by EE, in association with normalization of biliary excretion of glutathione. Western blot studies indicate that EE decreased the expression of multidrug resistance-associated protein 2 (Mrp2) by 41% and increased that of Mrp3 by 200%, whereas SL only affected Mrp2 expression (+60%) with respect to controls. The EE+SL group showed increased levels of Mrp2 and Mrp3 to the same extent as that registered for the individual treatments. Real-time polymerase chain reaction studies indicated that up-regulation of Mrp2 and Mrp3 by SL and EE, respectively, was at the transcriptional level. To estimate Mrp2 and Mrp3 activities, apical and basolateral excretion of acetaminophen glucuronide (APAP-glu), a common substrate for both transporters, was measured in the recirculating isolated perfused liver model. Biliary/perfusate excretion ratio was decreased in EE (-88%) and increased in SL (+36%) with respect to controls. Coadministration of rats with SL partially prevented (-53%) impairment induced by EE in this ratio. In conclusion, SL administration to EE-induced cholestatic rats counteracted the decrease in bile flow and biliary excretion of glutathione and APAP-glu, a model Mrp substrate, findings associated with up-regulation of Mrp2 expression.
Product Description
Supelco
Paracetamol β-D-glucuronide, analytical standard