MilliporeSigma
True moving bed electrophoresis using stepped electric field gradients.
Electrophoresis (2007-05-12)
Brian M Thome, Cornelius F Ivory
ABSTRACT
True moving bed electrophoresis has been shown to be an effective technique for the bench-scale separation of enantiomers, and it is desired to increase the maximum possible throughput attainable with the process by using electric field gradients. Homatropine enantiomer separations were performed and results using a stepped electric field gradient were compared to those using a traditional non-gradient separation. In order to accomplish this, a newly designed stator was constructed for use with the Vortex-Stabilized Electrophoresis Apparatus that has three sets of electrode housings, one set at both ends and one in the middle of the chamber. There were several problems related to the membranes used at the middle electrode. The dialysis membranes were permeable to the homatropine enantiomers, and while a switch to anion exchange membranes prevented the permeation of the homatropine, this caused a pH shift that interrupted binding to the hydroxypropyl-ss-cyclodextrin chiral selector. These problems prevented any meaningful data from being collected using homatropine enantiomers, and due to this, a proof of concept study was conducted using two bovine proteins. The separations using fluorescein-labeled BSA and bovine hemoglobin showed that a 63% increase in the maximum processing rate was attainable. The maximum throughput using the non-gradient process was 30.6 mg/h and the maximum was 50.0 mg/h using an electric field gradient that was 10% lower than the non-gradient field in section II and 10% higher in section III.
Product Description
Homatropine hydrobromide, European Pharmacopoeia (EP) Reference Standard