MilliporeSigma
  • De novo asymmetric syntheses of C-4-substituted sugars via an iterative dihydroxylation strategy.
De novo asymmetric syntheses of C-4-substituted sugars via an iterative dihydroxylation strategy.
Carbohydrate research (2006-04-18)
Md Moinuddin Ahmed, George A O'Doherty
ABSTRACT
A short and highly efficient route to various C-4 substituted sugar lactones has been developed. The key to the overall transformation is the sequential osmium-catalyzed dihydroxylation reaction of substituted 2,4-dienoates and an allylic substitution at the C-4 position. When the Sharpless AD-mix procedure is used in a matched sense for the second dihydroxylation reaction, it results in an exceedingly diastereo- and enantioselective synthesis of several C-4-substituted sugars.
Sigma-Aldrich
L-Galactono-1,4-lactone, ≥95.0% (GC)