MilliporeSigma
  • Effect of beclomethasone dipropionate and dexamethasone isonicotinate on lung function, bronchoalveolar lavage fluid cytology, and transcription factor expression in airways of horses with recurrent airway obstruction.
Effect of beclomethasone dipropionate and dexamethasone isonicotinate on lung function, bronchoalveolar lavage fluid cytology, and transcription factor expression in airways of horses with recurrent airway obstruction.
Journal of veterinary internal medicine (2006-04-06)
Laurent L Couëtil, Tatiana Art, Brieuc de Moffarts, Martine Becker, Dorothée Mélotte, Fabrice Jaspar, Fabrice Bureau, Pierre Lekeux
ABSTRACT
Glucocorticoid (GC) therapy is recognized to be effective for the treatment of recurrent airway obstruction (RAO) in horses. Anti-inflammatory properties of GC are thought to be mediated by suppression of inflammatory gene expression via inhibition of transcription factors such as nuclear factor-kappaB (NF-kappaB) and activator protein-1 (AP-1). The purpose of this study was to evaluate the effect of low-dose inhaled beclomethasone dipropionate and injectable dexamethasone 21-isonicotinate on clinical signs, pulmonary function, airway cytology, and activity of NF-kappaB and AP-1 in bronchial cells of RAO-affected horses. Seven horses with RAO were exposed to moldy hay until they developed airway obstruction on 3 separate occasions. In a crossover design, they were then treated with a placebo (injection on day 1), inhaled beclomethasone (500 microg q12h for 10 days), or dexamethasone (0.06 mg/kg, IM on day 1) and monitored for 10 days. Pulmonary function, bronchoalveolar lavage fluid cytology, and NF-kappaB and AP-1 activity in bronchial brushing cells were measured before (day 1) and after treatment (day 10). Treatment with beclomethasone resulted in significantly improved pulmonary function of RAO-affected horses compared with placebo and dexamethasone treatments. However, none of the treatments had an effect on bronchoalveolar lavage fluid cytology or NF-kappaB and AP-1 activity. These findings reveal that, in a model of severe RAO, the benefits of low-dose inhaled beclomethasone on pulmonary function are not accompanied by a decrease in airway inflammatory cells or a suppression of transcription factors NF-kappaB and AP-1 DNA-binding activity.
Product Description
Dexamethasone isonicotinate, European Pharmacopoeia (EP) Reference Standard
Dexamethasone isonicotinate for impurity C identification, European Pharmacopoeia (EP) Reference Standard