MilliporeSigma
  • A biomimetic approach to the rocaglamides employing photogeneration of oxidopyryliums derived from 3-hydroxyflavones.
A biomimetic approach to the rocaglamides employing photogeneration of oxidopyryliums derived from 3-hydroxyflavones.
Journal of the American Chemical Society (2004-10-21)
Baudouin Gerard, Guilford Jones Ii, John A Porco
ABSTRACT
A unified biomimetic approach to the aglain-forbaglin-rocaglamide classes of natural products is reported. The approach involves photogeneration of oxidopyryliums via excited-state intramolecular proton transfer (ESIPT) of 3-hydroxyflavones followed by [3+2] dipolar cycloaddition to the aglain core. An alpha-ketol (acyloin) rearrangement was employed to transform the aglain core to the rocaglamide framework. This approach was successfully used for the synthesis of the natural product (+/-)-methyl rocaglate.
Sigma-Aldrich
Rocaglamide, ≥96% (HPLC)