MilliporeSigma
  • Multi-detector row CT: effect of iodine dose reduction on hepatic and vascular enhancement.
Multi-detector row CT: effect of iodine dose reduction on hepatic and vascular enhancement.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin (2004-04-17)
J E Roos, L M Desbiolles, D Weishaupt, S Wildermuth, P R Hilfiker, B Marincek, T Boehm
ABSTRACT
To evaluate the impact of different iodine concentrations of intravenous contrast agent on hepatic and vascular enhancement during arterial and porto-venous phase imaging using a 4-channel multi-detector row CT (MDCT). One hundred consecutive patients referred for triphasic abdominal MDCT were randomly assigned into four groups receiving different iodine concentration (200, 250, 300 or 350 mg/ml). Non-contrast, arterial, and porto-venous phase 4-channel MDCT imaging was performed (VolumeZoom, Siemens, Germany). A fixed volume of 150 ml intravenous contrast agent at a rate of 3 ml/s was injected using an automatic bolus-tracking system (Care Bolus, Siemens, Erlangen). Hepatic and vascular enhancement values were measured over time and non-contrast values were subtracted in order to compute arterial and porto-venous mean hepatic (MHE) and mean aortic (MAE) enhancement for each group. Mean change of enhancement > 80 HU for the aorta and > 40 HU for the liver during porto-venous phase imaging was considered as sufficient enhancement. All groups achieved sufficient vascular enhancement during arterial phase imaging; MAE with 350 mg/ml (222 HU) and 300 mg/ml (213HU) was significantly better than with 250 mg (196HU) and 200 mg/ml (169 HU), whereas MHE showed no statistically significant difference between the groups (range 16 - 25 HU). Porto-venous MHE showed increased enhancement with larger concentrations, with significant differences among the groups. Only the higher concentration groups (350 mg/ml und 300 mg/ml) fulfilled in every individual the guidelines for sufficient porto-venous MHE. In the lower concentration groups, 8 patients with 200 mg/ml and 3 patients with 250 mg/ml showed enhancement values below the required minimum. A decrease in iodine contrast agent down to 200 mg/ml concentration is only tenable for propose of vascular aortic and hepatic arterial enhancement, whereas hepatic porto-venous phase imaging still requires concentrations at or above the level of 300 mg/ml.
Product Description
Iopentol, European Pharmacopoeia (EP) Reference Standard
USP
Iodixanol Related Compound D, United States Pharmacopeia (USP) Reference Standard