MilliporeSigma
Iridoids as allelochemicals and DNA polymerase inhibitors.
Journal of natural products (2004-03-27)
Carlos R Pungitore, Maximiliano Juri Ayub, Matías García, Eduardo J Borkowski, Marta E Sosa, Gladys Ciuffo, Oscar S Giordano, Carlos E Tonn
ABSTRACT
Growth inhibitory activities and nutritional indices of catalpol (1), 8-O-acetylharpagide (2), and harpagide (3) were determinated in larvae and adults of Tribolium castaneum, respectively. Compound 1 produced a series of allelochemical effects probably related with the DNA synthesis. This iridoid possessed the highest inhibitory activity against DNA polymerase. Molecular orbital calculations suggest that a pi-pi charge transfer recognition model could explain the action of iridioids toward nucleic acid synthesis.
Sigma-Aldrich
8-Acetylharpagide, ≥85% (LC/MS-ELSD)