MilliporeSigma
[Compounds from roots of Chirita fimbrisepala Hand.-Mazz].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2003-01-15)
L D Zhou, J G Yu, J Guo, S L Yang
ABSTRACT
To study the chemical constituents the roots of Chirita fimbrisepala. The constituents were extracted with solvent, separated and purified with chromatographic methods, identified by NMR, MS, UV, IR and physical-chemical constants. Three flavonoids mahuangchiside(I), hispidulin (II) and kaempferol(III) were isolated with daucosteral(IV). I is a new compound elucidated as hispidulin-7-O-beta-D-xylopyranosyl-(1-->2)-beta-D-xylopyranoside, named mahuangchiside, II and III were isolated for the first time from the family Gesneriaceae, and IV was isolated for the first time from the genus Chirita.
Sigma-Aldrich
Hispidulin, ≥98% (HPLC)