MilliporeSigma
  • Inhibition of HIV-1 integrase by galloyl glucoses from Terminalia chebula and flavonol glycoside gallates from Euphorbia pekinensis.
Inhibition of HIV-1 integrase by galloyl glucoses from Terminalia chebula and flavonol glycoside gallates from Euphorbia pekinensis.
Planta medica (2002-06-12)
Mi-Jeong Ahn, Chul Young Kim, Ji Suk Lee, Tae Gyun Kim, Seung Hee Kim, Chong-Kyo Lee, Bo-Bin Lee, Cha-Gyun Shin, Hoon Huh, Jinwoong Kim
ABSTRACT
The bioassay-directed isolation of Terminalia chebula fruits afforded four human immunodeficiency virus type 1 (HIV-1) integrase inhibitors, gallic acid ( 1) and three galloyl glucoses ( 2 - 4). In addition, four flavonol glycoside gallates ( 5 - 8) from Euphorbia pekinensis containing the galloyl moiety also showed the inhibitory activity at a level comparable to those of 2 - 4. By comparison with the activities of the compounds not bearing this moiety, it is proposed that the galloyl moiety plays a major role for inhibition against the 3'-processing of HIV-1 integrase of these compounds.
Product Description
Supelco
Chebulagic acid, analytical standard