MilliporeSigma
  • The metabolism of norethandrolone in the horse: characterization of 16-, 20- and 21-oxygenated metabolites by gas chromatography/mass spectrometry.
The metabolism of norethandrolone in the horse: characterization of 16-, 20- and 21-oxygenated metabolites by gas chromatography/mass spectrometry.
Journal of mass spectrometry : JMS (2001-04-05)
A R McKinney, D D Ridley, C J Suann
ABSTRACT
After oral administration to a thoroughbred gelding, the anabolic steroid norethandrolone was converted into a complex mixture of oxygenated metabolites. These metabolites were extracted from the urine, deconjugated by methanolysis and converted to their O-methyloxime trimethylsilyl derivatives. Gas chromatographic/mass spectrometric analysis indicated the major metabolites to be 19-norpregnane-3,16,17-triols, 19-norpregnane-3,17,20-triols and 3,17-dihydroxy-19-norpregnan-21-oic acids. Some minor metabolites were also detected.