MilliporeSigma
  • Tiaprofenic acid-photosensitized damage to nucleic acids: a mechanistic study using complementary in vitro approaches.
Tiaprofenic acid-photosensitized damage to nucleic acids: a mechanistic study using complementary in vitro approaches.
Photochemistry and photobiology (2000-05-20)
C Agapakis-Caussé, F Boscá, J V Castell, D Hernández, M L Marín, L Marrot, M A Miranda
ABSTRACT
In order to determine whether or not tiaprofenic acid (TPA) could cause cellular DNA damage, human fibroblasts were irradiated in the presence of the drug and subsequently examined by means of the comet assay. This led to the observation that TPA actually sensitizes cellular DNA to the subsequent irradiation. When TPA was irradiated in the presence of supercoiled plasmid DNA, it produced large amounts of single-strand breaks (SSB); this is consistent with the effects observed on cellular genomic DNA by the comet assay. More importantly, low concentrations of TPA, unable to produce direct SSB, caused photo-oxidative damage to DNA as revealed by the use of excision-repair enzymes. The fact that TPA-irradiated DNA was a substrate of formamidopyrimidine glycosylase as well as endonuclease III revealed that both purine and pyrimidine bases were oxidized. This was further supported by the TPA-photosensitized oxidation of 2'-deoxyguanosine which led to a product mixture characteristic of mixed type-I/II mechanisms. Thymidine was less reactive under similar conditions, but it also decomposed to give a typical type-I product pattern. Accordingly, the TPA triplet was quenched by the two nucleosides with clearly different rate constants (10(8) vs 10(7) M-1 s-1, respectively). As cellular RNA also contains oxidizable bases, it could be the target of similar processes, thus interfering with the biosynthesis of proteins by the cells. Extraction of total RNA from TPA-irradiated human fibroblasts, followed by gel electrophoresis and PCR analysis, confirmed this hypothesis. Finally, photosensitization experiments with Saccharomyces cerevisiae showed that, in spite of an efficient drug-yeast interaction leading to cytotoxicity, neither intergenic recombination nor gene conversion took place. Thus, while TPA-photosensitized damage to nucleic acids can result in genotoxicity, the risk of mutagenicity does not appear to be significant.
Product Description
Tiaprofenic acid, European Pharmacopoeia (EP) Reference Standard