MilliporeSigma
  • Silylium ion-promoted dehydrogenative cyclization: synthesis of silicon-containing compounds derived from alkynes.
Silylium ion-promoted dehydrogenative cyclization: synthesis of silicon-containing compounds derived from alkynes.
Chemical communications (Cambridge, England) (2014-05-16)
Hidekazu Arii, Takashi Kurihara, Kunio Mochida, Takayuki Kawashima
ABSTRACT
Treatment of dialkylbenzylsilane (1) with trityl tetrakis(pentafluorophenyl)borate (TPFPB) in the presence of terminal or internal alkynes (3) and 2,6-di-tert-butyl-4-methylpyridine gave the corresponding 1,2-dihydro-2-silanaphthalene derivatives (4) in 34-82% yields.
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylpyridine, 98%