MilliporeSigma
  • Pleiotrophin commits human bone marrow mesenchymal stromal cells towards hypertrophy during chondrogenesis.
Pleiotrophin commits human bone marrow mesenchymal stromal cells towards hypertrophy during chondrogenesis.
PloS one (2014-02-12)
Thibault Bouderlique, Emilie Henault, Angelique Lebouvier, Guilhem Frescaline, Phillipe Bierling, Helene Rouard, José Courty, Patricia Albanese, Nathalie Chevallier
ABSTRACT
Pleiotrophin (PTN) is a growth factor present in the extracellular matrix of the growth plate during bone development and in the callus during bone healing. Bone healing is a complicated process that recapitulates endochondral bone development and involves many cell types. Among those cells, mesenchymal stromal cells (MSC) are able to differentiate toward chondrogenic and osteoblastic lineages. We aimed to determine PTN effects on differentiation properties of human bone marrow stromal cells (hBMSC) under chondrogenic induction using histological analysis and quantitative reverse transcription polymerase chain reaction. PTN dramatically potentiated chondrogenic differentiation as indicated by a strong increase of collagen 2 protein, and cartilage-related gene expression. Moreover, PTN increased transcription of hypertrophic chondrocyte markers such as MMP13, collagen 10 and alkaline phosphatase and enhanced calcification and the content of collagen 10 protein. These effects are dependent on PTN receptors signaling and PI3 K pathway activation. These data suggest a new role of PTN in bone regeneration as an inducer of hypertrophy during chondrogenic differentiation of hBMSC.
Sigma-Aldrich
Anti-Collagen Type II Antibody, clone 6B3, clone 6B3, Chemicon®, from mouse