MilliporeSigma
  • Direct chiral resolution of malic acid in apple juice by ligand-exchange capillary electrophoresis using copper(II)-L-tartaric acid as a chiral selector.
Direct chiral resolution of malic acid in apple juice by ligand-exchange capillary electrophoresis using copper(II)-L-tartaric acid as a chiral selector.
Electrophoresis (2001-10-09)
S Kodama, A Yamamoto, A Matsunaga, T Soga, K Hayakawa
ABSTRACT
Chiral resolution of native DL-malic acid was achieved by ligand-exchange capillary electrophoresis using copper(II)-L-tartrate as a chiral selector. Factors affecting chiral resolution, migration time, and peak area of malic acid were studied. The running conditions for optimum separation of malic acid were found to be 1 mM copper(II) sulfate-1 mM L-tartrate (pH 5.1) with an effective voltage of -20 kV at 30 degrees C, using direct detection at 280 nm, and resolution (Rs) of racemic malic acid was approximately 4. With this system, D- and L-malic acids in apple juice were analyzed successfully.
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%