MilliporeSigma
  • Pressure dependent product formation in the photochemically initiated allyl + allyl reaction.
Pressure dependent product formation in the photochemically initiated allyl + allyl reaction.
Molecules (Basel, Switzerland) (2013-11-07)
Lars Seidel, Karlheinz Hoyermann, Fabian Mauß, Jörg Nothdurft, Thomas Zeuch
ABSTRACT
Photochemically driven reactions involving unsaturated radicals produce a thick global layer of organic haze on Titan, Saturn's largest moon. The allyl radical self-reaction is an example for this type of chemistry and was examined at room temperature from an experimental and kinetic modelling perspective. The experiments were performed in a static reactor with a volume of 5 L under wall free conditions. The allyl radicals were produced from laser flash photolysis of three different precursors allyl bromide (C3H5Br), allyl chloride (C3H5Cl), and 1,5-hexadiene (CH2CH(CH2)2CHCH2) at 193 nm. Stable products were identified by their characteristic vibrational modes and quantified using FTIR spectroscopy. In addition to the (re-) combination pathway C3H5+C3H5 → C6H10 we found at low pressures around 1 mbar the highest final product yields for allene and propene for the precursor C3H5Br. A kinetic analysis indicates that the end product formation is influenced by specific reaction kinetics of photochemically activated allyl radicals. Above 10 mbar the (re-) combination pathway becomes dominant. These findings exemplify the specificities of reaction kinetics involving chemically activated species, which for certain conditions cannot be simply deduced from combustion kinetics or atmospheric chemistry on Earth.
Sigma-Aldrich
Allyl chloride, ReagentPlus®, 99%
Pricing and availability is not currently available.