MilliporeSigma
Synthesis of a water-soluble protein cross-linking reagent.
Biological chemistry Hoppe-Seyler (1990-09-01)
U M Kremer
ABSTRACT
2,4-Difluoro-5-nitrobenzenesulfonic acid has been synthesized by the sulfonation of 2,4-difluoronitrobenzene, and precipitated with KCl as the potassium sulfonate. The structure was confirmed by chemical and spectroscopic methods (IR, 1H-NMR, 13C-NMR, 19F-NMR, UV, MS and ultimate organic analysis). Lysozyme was cross-linked with the potassium sulfonate and with 1,5-difluoro-2,4-dinitrobenzene. The products were analysed by SDS-PAGE and compared. The cross-linking conditions were optimized.
Sigma-Aldrich
2,4-Difluoronitrobenzene, 99%