MilliporeSigma
  • Synthesis and in vitro toxicity of hydroxylamine metabolites of sulfonamides.
Synthesis and in vitro toxicity of hydroxylamine metabolites of sulfonamides.
The Journal of pharmacology and experimental therapeutics (1988-02-01)
M J Rieder, J Uetrecht, N H Shear, S P Spielberg
ABSTRACT
Among the most serious side effects of sulfonamides are hypersensitivity reactions, the pathogenesis of which has been suggested to be mediated by reactive metabolites. We have previously demonstrated dose-related covalent binding and toxicity of reactive intermediates of sulfonamides generated by a murine hepatic microsomal activating system. We hypothesized that hydroxylamine (H/A) metabolites might be likely candidates for mediating such toxicity; accordingly, we synthesized chemically the H/As of sulfadiazine and sulfamethoxazole. Synthesis was performed using 4-nitrobenzenesulfonyl chloride and either 2-aminopyrimidine or 3-amino-5-methylisoxazole, respectively, as starting materials. The resulting nitro derivatives were reduced to the corresponding H/A with hydrogen in the presence of a poisoned platinum catalyst. After synthesis and purification, toxicity of the H/As to lymphocytes of normal volunteers was evaluated using three cytotoxicity assays: 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl tetrazolium bromide dye conversion, trypan blue dye exclusion and propidium iodide dye exclusion. The H/As of sulfadiazine and sulfamethoxazole displayed dose-related toxicity. 1.6 mM sulfadiazine H/A produced 82% cell death, whereas 400 microM sulfamethoxazole H/A produced 62% cell death; the parent sulfonamides were not toxic to cells. The toxicity of sulfamethoxazole H/A was decreased by coincubation with glutathione or N-acetylcysteine; there was a 47% decrease in toxicity when coincubated with 100 microM glutathione, whereas there was a 55% decrease displayed when coincubation was done with 500 microM N-acetylcysteine. H/A metabolites of the sulfonamides or their nitroso derivatives, normally detoxified by conjugation to glutathione, may be the proximate toxins mediating sulfonamide hypersensitivity.
Sigma-Aldrich
4-Nitrobenzenesulfonyl chloride, 97%
Sigma-Aldrich
3-Amino-5-methylisoxazole, ≥97%