MilliporeSigma
  • Benzamide bioisosteres incorporating dihydroheteroazole substructures: EPC synthesis and SAR leading to a selective dopamine D4 receptor partial agonist (FAUC 179).
Benzamide bioisosteres incorporating dihydroheteroazole substructures: EPC synthesis and SAR leading to a selective dopamine D4 receptor partial agonist (FAUC 179).
Bioorganic & medicinal chemistry letters (2001-09-11)
J Einsiedel, H Hübner, P Gmeiner
ABSTRACT
Conformationally restricted benzamide bioisosteres were investigated when the chiral phenyldihydroimidazole derivative 4e (FAUC 179) showed strong and highly selective dopamine D4 receptor binding (K(i)high=0.95nM). Mitogenesis experiments indicated partial agonist properties (42%). EPC syntheses of the target compounds of type 4 were performed starting from alpha-amino acids.
Sigma-Aldrich
Methyl benzimidate hydrochloride, 97%