MilliporeSigma
  • The final step in the phage infection cycle: the Rz and Rz1 lysis proteins link the inner and outer membranes.
The final step in the phage infection cycle: the Rz and Rz1 lysis proteins link the inner and outer membranes.
Molecular microbiology (2008-08-21)
Joel Berry, Elizabeth J Summer, Douglas K Struck, Ryland Young
ABSTRACT
Bacteriophage lambda has four adjacent genes -S, R, Rz and Rz1- dedicated to host cell lysis. While S, encoding the holin and antiholin, and R, encoding the endolysin, have been intensively studied, the products of Rz and Rz1 have not been characterized at either the structural or functional levels. Rz1 is an outer membrane lipoprotein and our results indicate that Rz is a type II signal anchor protein. Here we present evidence that an Rz-Rz1 complex that spans the periplasm carries out the final step in the process of host lysis. These results are discussed in terms of a model where endolysin-mediated degradation of the cell wall is a prerequisite for conformational changes in the Rz-Rz1 complex leading to the juxtaposition and fusion of the IM and OM. Fusion of the two membranes removes the last physical barrier to efficient release of progeny virions.
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for use with bacterial cell extracts, lyophilized powder
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution