MilliporeSigma
  • Efficient solvent- and metal-free Sonogashira protocol catalysed by 1,4-diazabicyclo(2.2.2) octane (DABCO).
Efficient solvent- and metal-free Sonogashira protocol catalysed by 1,4-diazabicyclo(2.2.2) octane (DABCO).
Organic & biomolecular chemistry (2009-04-04)
Rafael Luque, Duncan J Macquarrie
ABSTRACT
An efficient metal-free Sonogashira coupling protocol catalysed by DABCO is reported, where very good conversions and selectivities to the cross-coupling product were obtained under mild reaction conditions. The reported solvent-, phosphane- and metal-free protocol, that uses a cheap base as catalyst, considerably improves the green credentials of the reaction.
Sigma-Aldrich
3-Iodobenzotrifluoride, 98%