MilliporeSigma
  • Initial structure-activity relationship studies of a novel series of pyrrolo[1,2-a]pyrimid-7-ones as GnRH receptor antagonists.
Initial structure-activity relationship studies of a novel series of pyrrolo[1,2-a]pyrimid-7-ones as GnRH receptor antagonists.
Bioorganic & medicinal chemistry letters (2002-01-30)
Yun-Fei Zhu, R Scott Struthers, Patrick J Connors, Yinghong Gao, Timothy D Gross, John Saunders, Keith Wilcoxen, Greg J Reinhart, Nicholas Ling, Chen Chen
ABSTRACT
Initial SAR studies on 1-aminomethyl-2-aryl-3-cyano-pyrrolo[1,2-a]pyrimid-7-one-6-carboxylates as human GnRH receptor antagonists were discussed. 2-(2-Methylaminoethyl)pyridine was discovered to be a key feature for generating active compounds. The best compound from the series had 25 nM (K(i)) binding affinity to human GnRH receptor.