MilliporeSigma
  • Regulation of the MAFF transcription factor by proinflammatory cytokines in myometrial cells.
Regulation of the MAFF transcription factor by proinflammatory cytokines in myometrial cells.
Biology of reproduction (2005-12-24)
Wael Massrieh, Anna Derjuga, Florence Doualla-Bell, Chun-Ying Ku, Barbara M Sanborn, Volker Blank
ABSTRACT
The MAF (proto-)oncogene family of basic-leucine zipper transcription factors plays crucial roles in the control of mammalian gene expression and development. Here we analyzed the regulation of the human MAFF gene, coding for a small MAF transcription factor, in uterine smooth muscle cells. We found that MAFF transcript levels are induced by proinflammatory cytokines in PHM1-31 myometrial cells. We observed an important induction by interleukin 1 beta (IL1B) and a weaker upregulation by tumor necrosis factor (TNF), whereas interleukin 6 (IL6) treatment had no effect. Time course experiments revealed a rapid induction of MAFF transcripts within 30 min following IL1B treatment. The presence of actinomycin D inhibited the upregulation, suggesting that regulation of MAFF mRNA levels occurs at the transcriptional level. We generated a MAFF-specific antiserum and determined that MAFF protein was also induced by TNF and IL1B in PHM1-31 cells. In contrast, it was particularly interesting that the transcript and protein levels of the highly homologous MAFG and MAFK genes are not modulated by these cytokines. Our results suggest a possible specific role for MAFF in proinflammatory cytokine-mediated control of myometrial gene expression and provide the first link between a small MAF transcription factor and the inflammatory response.
Sigma-Aldrich
Anti-MAFF antibody produced in rabbit, IgG fraction of antiserum