MilliporeSigma
  • Lead(II) thiocyanate complexes with bibracchial lariat ethers: an X-ray and DFT study.
Lead(II) thiocyanate complexes with bibracchial lariat ethers: an X-ray and DFT study.
Inorganic chemistry (2005-03-29)
Carlos Platas-Iglesias, David Esteban-Gómez, Teresa Enríquez-Pérez, Fernando Avecilla, Andrés de Blas, Teresa Rodríguez-Blas
ABSTRACT
Compounds of formula [Pb(L2)(NCS)2] (1) and [Pb(L4)(SCN)2] (2) (where L2 is the lariat crown ether N,N'-bis(3-aminobenzyl)-4,13-diaza-18-crown-6 and L4 is the Schiff-base lariat crown ether N,N'-bis(3-(salicylaldimino)benzyl)-4,13-diaza-18-crown-6) were isolated and structurally characterized by X-ray diffraction analyses. The X-ray crystal structures of both compounds show the metal ion coordinated to the six donor atoms of the crown moiety, leaving the corresponding pendant arms uncoordinated. The coordination sphere of lead(II) is completed by two thiocyanate groups that coordinate either through their nitrogen (1) or sulfur (2) atoms. The organic receptor adopts a syn conformation in 1, while in 2 it shows an anti conformation. To rationalize these unexpected different conformations of the L2 and L4 receptors in compounds 1 and 2, as well as the different binding modes found for the thiocyanate ligands, we have carried out theoretical calculations at the DFT (B3LYP) level. These calculations predict the syn conformation being the most stable in both 1 and 2 complexes. So, the anti conformation found for 2 in the solid state is tentatively attributed to the presence of intermolecular pi-pi interactions between phenol rings, for which the dihedral angle between the least-squares planes of both rings amounts to 2.6 degrees and the distance between the center of both rings is 3.766 A. On the other hand, the analysis of the electronic structure has revealed that the Pb-ligand bonds present highly ionic character in this family of compounds. They also suggest a greater transfer of electron density from the NCS- ligands when they coordinate through the less electronegative S atom. The Pb-SCN covalent bond formation mainly occurs due to an effective overlap of the occupied 3p z orbitals of the S atoms and the unoccupied 6p z AO of the Pb atom, while the Pb-NCS bonding interaction is primarily due to the overlap of the 6s and 7s AO of Pb with sp(1.10) hybrids of the N donor atoms. Our electronic structure calculations can rationalize the different coordination of the thiocyanate groups in compounds 1 and 2: the simultaneous formation of two Pb-SCN bonds is more favorable for S-Pb-S angles close to 180 degrees , for which the overlap between the occupied 3p z orbitals of the S atoms and the unoccupied 6 pz AO of the Pb atom is maximized.