MilliporeSigma
  • Binding of 5-methoxypsoralen to blood fractions. Influence of albumin and free fatty acids: an in vitro study.
Binding of 5-methoxypsoralen to blood fractions. Influence of albumin and free fatty acids: an in vitro study.
Skin pharmacology : the official journal of the Skin Pharmacology Society (1993-01-01)
P Muret, S Makki, S Urien, P Agache, P Bechtel, J P Tillement
ABSTRACT
The binding of 5-methoxypsoralen (5-MOP) to human serum and blood fractions was studied by equilibrium dialysis associated to high-performance liquid chromatography. 5-MOP serum binding was 95% and kept constant in the range of therapeutic concentrations. Albumin was the main binding protein with one class of binding sites (n = 2.05) and with a moderate affinity constant (Ka = 10,270 M-1). Free fatty acids (FFA) enhanced 5-MOP binding to albumin. Binding to other proteins, alpha 1-acid glycoprotein, gamma-globulins and lipoproteins (LDL, HDL, VLDL) also occurred, but was negligible. 5-MOP bound to red blood cells (RBCs) with a binding index (ratio of intra- to extra-globular 5-MOP concentrations) of 3.84 and 1.48 in buffer and plasma, respectively. 5-MOP distribution in blood was simulated. The free fraction of 5-MOP in blood could be increased in hypoalbuminemia, unchanged in acute inflammation, and decreased by FFA.
Sigma-Aldrich
Globulins Cohn fraction II, III human, lyophilized powder
Sigma-Aldrich
Globulins Cohn fraction IV-4 human