MilliporeSigma
  • Synthesis and antiinflammatory activity of 2,3-dihydro-1,4-benzodioxin methyl carboxylic acids.
Synthesis and antiinflammatory activity of 2,3-dihydro-1,4-benzodioxin methyl carboxylic acids.
Farmaco (Societa chimica italiana : 1989) (1996-03-01)
M T Vázquez, G Rosell, M D Pujol
ABSTRACT
Synthesis and antiinflammatory properties of new carboxylic acids containing the 2,3-dihydro-1,4-benzodioxin subunit are described. The 2-(2,3-dihydro-1, 4-benzodioxin-6-yl)acetic acid was of comparable potency to Ibuprofen, in carrageenan induced rat paw edema assay.