MilliporeSigma
  • Bevacizumab plus radiotherapy-temozolomide for newly diagnosed glioblastoma.
Bevacizumab plus radiotherapy-temozolomide for newly diagnosed glioblastoma.
The New England journal of medicine (2014-02-21)
Olivier L Chinot, Wolfgang Wick, Warren Mason, Roger Henriksson, Frank Saran, Ryo Nishikawa, Antoine F Carpentier, Khe Hoang-Xuan, Petr Kavan, Dana Cernea, Alba A Brandes, Magalie Hilton, Lauren Abrey, Timothy Cloughesy
ABSTRACT
Standard therapy for newly diagnosed glioblastoma is radiotherapy plus temozolomide. In this phase 3 study, we evaluated the effect of the addition of bevacizumab to radiotherapy-temozolomide for the treatment of newly diagnosed glioblastoma. We randomly assigned patients with supratentorial glioblastoma to receive intravenous bevacizumab (10 mg per kilogram of body weight every 2 weeks) or placebo, plus radiotherapy (2 Gy 5 days a week; maximum, 60 Gy) and oral temozolomide (75 mg per square meter of body-surface area per day) for 6 weeks. After a 28-day treatment break, maintenance bevacizumab (10 mg per kilogram intravenously every 2 weeks) or placebo, plus temozolomide (150 to 200 mg per square meter per day for 5 days), was continued for six 4-week cycles, followed by bevacizumab monotherapy (15 mg per kilogram intravenously every 3 weeks) or placebo until the disease progressed or unacceptable toxic effects developed. The coprimary end points were investigator-assessed progression-free survival and overall survival. A total of 458 patients were assigned to the bevacizumab group, and 463 patients to the placebo group. The median progression-free survival was longer in the bevacizumab group than in the placebo group (10.6 months vs. 6.2 months; stratified hazard ratio for progression or death, 0.64; 95% confidence interval [CI], 0.55 to 0.74; P<0.001). The benefit with respect to progression-free survival was observed across subgroups. Overall survival did not differ significantly between groups (stratified hazard ratio for death, 0.88; 95% CI, 0.76 to 1.02; P=0.10). The respective overall survival rates with bevacizumab and placebo were 72.4% and 66.3% at 1 year (P=0.049) and 33.9% and 30.1% at 2 years (P=0.24). Baseline health-related quality of life and performance status were maintained longer in the bevacizumab group, and the glucocorticoid requirement was lower. More patients in the bevacizumab group than in the placebo group had grade 3 or higher adverse events (66.8% vs. 51.3%) and grade 3 or higher adverse events often associated with bevacizumab (32.5% vs. 15.8%). The addition of bevacizumab to radiotherapy-temozolomide did not improve survival in patients with glioblastoma. Improved progression-free survival and maintenance of baseline quality of life and performance status were observed with bevacizumab; however, the rate of adverse events was higher with bevacizumab than with placebo. (Funded by F. Hoffmann-La Roche; ClinicalTrials.gov number, NCT00943826.).
Product Description
Supelco
Temozolomide, VETRANAL®, analytical standard
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)