MilliporeSigma
  • Intraoperative anaphylaxis to sugammadex and a protocol for intradermal skin testing.
Intraoperative anaphylaxis to sugammadex and a protocol for intradermal skin testing.
Anaesthesia and intensive care (2014-01-30)
P H M Sadleir, T Russell, R C Clarke, E Maycock, P R Platt
ABSTRACT
Sugammadex is a selective binding agent for aminosteroid neuromuscular blockers whose use is increasing in anaesthetic practice. We present three cases of severe anaphylaxis coincident with sugammadex administration. Subsequent intradermal testing confirmed sugammadex as the triggering agent, with all patients having positive skin responses to a 1:100 dilution of the standard 100 mg/ml solution and two out of three having a positive response to a 1:1000 dilution. As all patients were administered sugammadex to reverse neuromuscular blockade with rocuronium, we considered that sugammadex-rocuronium complexes were a potential unique allergen. In the two patients who were additionally tested with a rocuronium-sugammadex (3.6:1 molecular ratio) mixture, the wheal-and-flare response was significantly attenuated.
Product Description
Rocuronium for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rocuronium bromide, ≥97% (perchloric acid titration)