MilliporeSigma
  • Vitamin D during pregnancy and infancy and infant serum 25-hydroxyvitamin D concentration.
Vitamin D during pregnancy and infancy and infant serum 25-hydroxyvitamin D concentration.
Pediatrics (2013-12-18)
Cameron C Grant, Alistair W Stewart, Robert Scragg, Tania Milne, Judy Rowden, Alec Ekeroma, Clare Wall, Edwin A Mitchell, Sue Crengle, Adrian Trenholme, Julian Crane, Carlos A Camargo
ABSTRACT
To determine the vitamin D dose necessary to achieve serum 25-hydroxyvitamin D (25(OH)D) concentration ≥ 20 ng/mL during infancy. A randomized, double-blind, placebo-controlled trial in New Zealand. Pregnant mothers, from 27 weeks' gestation to birth, and then their infants, from birth to age 6 months, were randomly assigned to 1 of 3 mother/infant groups: placebo/placebo, vitamin D3 1000/400 IU, or vitamin D3 2000/800 IU. Serum 25(OH)D and calcium concentrations were measured at enrollment, 36 weeks' gestation, in cord blood, and in infants at 2, 4, and 6 months of age. Two-hundred-and-sixty pregnant women were randomized. At enrollment, the proportions with serum 25(OH)D ≥ 20 ng/mL for placebo, lower-dose, and higher-dose groups were 54%, 64%, and 55%, respectively. The proportion with 25(OH)D ≥ 20 ng/mL was larger in both intervention groups at 36 weeks' gestation (50%, 91%, 89%, P < .001). In comparison with placebo, the proportion of infants with 25(OH)D ≥ 20 ng/mL was larger in both intervention groups to age 4 months: cord blood (22%, 72%, 71%, P < .001), 2 months (50%, 82%, 92%, P < .001), and 4 months (66%, 87%, 87%, P = .004), but only in the higher-dose group at age 6 months (74%, 82%, 89%, P = .07; higher dose versus placebo P = .03, lower dose versus placebo P = .21). Daily vitamin D supplementation during pregnancy and then infancy with 1000/400 IU or 2000/800 IU increases the proportion of infants with 25(OH)D ≥ 20 ng/mL, with the higher dose sustaining this increase for longer.
Product Description
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material