MilliporeSigma
  • Cloning and characterization of canadine synthase involved in noscapine biosynthesis in opium poppy.
Cloning and characterization of canadine synthase involved in noscapine biosynthesis in opium poppy.
FEBS letters (2013-12-10)
Thu-Thuy T Dang, Peter J Facchini
ABSTRACT
Noscapine biosynthesis in opium poppy is thought to occur via N-methylcanadine, which would be produced through 9-O-methylation of (S)-scoulerine, methylenedioxy bridge formation on (S)-tetrahydrocolumbamine, and N-methylation of (S)-canadine. Only scoulerine 9-O-methyltransferase has been functionally characterized. We report the isolation and characterization of a cytochrome P450 (CYP719A21) from opium poppy that converts (S)-tetrahydrocolumbamine to (S)-canadine. Recombinant CYP719A21 displayed strict substrate specificity and high affinity (Km=4.63±0.71 μM) for (S)-tetrahydrocolumbamine. Virus-induced gene silencing of CYP719A21 caused a significant increase in (S)-tetrahydrocolumbamine accumulation and a corresponding decrease in the levels of putative downstream intermediates and noscapine in opium poppy plants.
Sigma-Aldrich
(S,R)-Noscapine, 97%