MilliporeSigma
  • Two analogues of fenarimol show curative activity in an experimental model of Chagas disease.
Two analogues of fenarimol show curative activity in an experimental model of Chagas disease.
Journal of medicinal chemistry (2013-12-07)
Martine Keenan, Jason H Chaplin, Paul W Alexander, Michael J Abbott, Wayne M Best, Andrea Khong, Adriana Botero, Catherine Perez, Scott Cornwall, R Andrew Thompson, Karen L White, David M Shackleford, Maria Koltun, Francis C K Chiu, Julia Morizzi, Eileen Ryan, Michael Campbell, Thomas W von Geldern, Ivan Scandale, Eric Chatelain, Susan A Charman
ABSTRACT
Chagas disease, caused by the protozoan parasite Trypanosoma cruzi (T. cruzi), is an increasing threat to global health. Available medicines were introduced over 40 years ago, have undesirable side effects, and give equivocal results of cure in the chronic stage of the disease. We report the development of two compounds, 6 and (S)-7, with PCR-confirmed curative activity in a mouse model of established T. cruzi infection after once daily oral dosing for 20 days at 20 mg/kg 6 and 10 mg/kg (S)-7. Compounds 6 and (S)-7 have potent in vitro activity, are noncytotoxic, show no adverse effects in vivo following repeat dosing, are prepared by a short synthetic route, and have druglike properties suitable for preclinical development.
Product Description
Supelco
Fenarimol, PESTANAL®, analytical standard