MilliporeSigma
  • Endogenous nitrated nucleotide is a key mediator of autophagy and innate defense against bacteria.
Endogenous nitrated nucleotide is a key mediator of autophagy and innate defense against bacteria.
Molecular cell (2013-11-26)
Chiaki Ito, Yohei Saito, Takashi Nozawa, Shigemoto Fujii, Tomohiro Sawa, Hirofumi Inoue, Tetsuro Matsunaga, Shahzada Khan, Soichiro Akashi, Ryota Hashimoto, Chihiro Aikawa, Eriko Takahashi, Hiroshi Sagara, Masaaki Komatsu, Keiji Tanaka, Takaaki Akaike, Ichiro Nakagawa, Hirokazu Arimoto
ABSTRACT
Autophagy is a cellular self-catabolic process wherein organelles, macromolecules, and invading microbes are sequestered in autophagosomes that fuse with lysosomes. In this study, we uncover the role of nitric oxide (NO) as a signaling molecule for autophagy induction via its downstream mediator, 8-nitroguanosine 3',5'-cyclic monophosphate (8-nitro-cGMP). We found that 8-nitro-cGMP-induced autophagy is mediated by Lys63-linked polyubiquitination and that endogenous 8-nitro-cGMP promotes autophagic exclusion of invading group A Streptococcus (GAS) from cells. 8-nitro-cGMP can modify Cys residues by S-guanylation of proteins. We showed that intracellular GAS is modified with S-guanylation extensively in autophagosomes-like vacuoles, suggesting the role of S-guanylation as a marker for selective autophagic degradation. This finding is supported by the fact that S-guanylated bacteria were selectively marked with polyubiquitin, a known molecular tag for selective transport to autophagosomes. These results collectively indicate that 8-nitro-cGMP plays a crucial role in cytoprotection during bacterial infections or inflammations via autophagy upregulation.
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Ubiquitin Antibody, Lys63-Specific, clone Apu3, rabbit monoclonal, clone Apu3, from rabbit
Sigma-Aldrich
Anti-Ubiquitin Antibody, Lys63-Specific, clone HWA4C4, clone HWA4C4, from mouse