MilliporeSigma
An unusual suspect in cocaine addiction.
Neuron (2013-11-26)
Yanhua H Huang, Oliver M Schlüter, Yan Dong
ABSTRACT
Development of drug addiction is extremely complex, but its initiation can be as simple as the flip-flop of glutamatergic receptor subtypes triggered by an "unusual" type of NMDA receptors, as suggested by Yuan et al. (2013) in this issue of Neuron.
Product Description
Supelco
Cocaine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cocaine hydrochloride solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis