MilliporeSigma
  • Site-selective displacement of tobramycin hydroxyls for preparation of antimicrobial cationic amphiphiles.
Site-selective displacement of tobramycin hydroxyls for preparation of antimicrobial cationic amphiphiles.
Organic letters (2013-11-15)
Yifat Berkov-Zrihen, Ido M Herzog, Mark Feldman, Micha Fridman
ABSTRACT
A short site-selective strategy for the activation and derivatization of alcohols of the clinically important aminoglycoside tobramycin is reported. The choice of amine protecting group affected the site-selective conversion of secondary alcohols of tobramycin into leaving groups. Temperature-dependent, chemoselective sequential nucleophilic displacements resulted in hetero- and homodithioether tobramycin-based cationic amphiphiles that demonstrated marked antimicrobial activity and impressive membrane selectivity.
Sigma-Aldrich
Tobramycin, Aminoglycoside antibiotic
Supelco
Tobramycin, Pharmaceutical Secondary Standard; Certified Reference Material