MilliporeSigma
  • Accidental spinal potassium chloride injection successfully treated with spinal lavage.
Accidental spinal potassium chloride injection successfully treated with spinal lavage.
Anaesthesia (2013-11-15)
J Dias, N Lages, A Marinho, L Maria, J Tinoco, D Vieira, C Correia
ABSTRACT
We describe the management of a 62-year-old man who developed severe pain, cramps, paraplegia and pulmonary oedema after the accidental administration of potassium chloride into the subarachnoid space. In addition to supportive treatment, we performed cerebrospinal fluid lavage with saline 0.9%. The patient recovered well without any permanent injury.
Product Description
Supelco
Bupivacaine hydrochloride monohydrate, analytical standard, for drug analysis
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.0%
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Supelco
Bupivacaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material