MilliporeSigma
  • A molecular basis for the association of the HLA-DRB1 locus, citrullination, and rheumatoid arthritis.
A molecular basis for the association of the HLA-DRB1 locus, citrullination, and rheumatoid arthritis.
The Journal of experimental medicine (2013-11-06)
Stephen W Scally, Jan Petersen, Soi Cheng Law, Nadine L Dudek, Hendrik J Nel, Khai Lee Loh, Lakshmi C Wijeyewickrema, Sidonia B G Eckle, Jurgen van Heemst, Robert N Pike, James McCluskey, Rene E Toes, Nicole L La Gruta, Anthony W Purcell, Hugh H Reid, Ranjeny Thomas, Jamie Rossjohn
ABSTRACT
Rheumatoid arthritis (RA) is strongly associated with the human leukocyte antigen (HLA)-DRB1 locus that possesses the shared susceptibility epitope (SE) and the citrullination of self-antigens. We show how citrullinated aggrecan and vimentin epitopes bind to HLA-DRB1*04:01/04. Citrulline was accommodated within the electropositive P4 pocket of HLA-DRB1*04:01/04, whereas the electronegative P4 pocket of the RA-resistant HLA-DRB1*04:02 allomorph interacted with arginine or citrulline-containing epitopes. Peptide elution studies revealed P4 arginine-containing peptides from HLA-DRB1*04:02, but not from HLA-DRB1*04:01/04. Citrullination altered protease susceptibility of vimentin, thereby generating self-epitopes that are presented to T cells in HLA-DRB1*04:01(+) individuals. Using HLA-II tetramers, we observed citrullinated vimentin- and aggrecan-specific CD4(+) T cells in the peripheral blood of HLA-DRB1*04:01(+) RA-affected and healthy individuals. In RA patients, autoreactive T cell numbers correlated with disease activity and were deficient in regulatory T cells relative to healthy individuals. These findings reshape our understanding of the association between citrullination, the HLA-DRB1 locus, and T cell autoreactivity in RA.
Sigma-Aldrich
L-Citrulline, ≥98% (TLC)