MilliporeSigma
  • Dynamic release and clearance of circulating microparticles during cardiac stress.
Dynamic release and clearance of circulating microparticles during cardiac stress.
Circulation research (2013-10-22)
Daniel Augustine, Lisa V Ayers, Eduardo Lima, Laura Newton, Adam J Lewandowski, Esther F Davis, Berne Ferry, Paul Leeson
ABSTRACT
Microparticles are cell-derived membrane vesicles, relevant to a range of biological responses and known to be elevated in cardiovascular disease. To investigate microparticle release during cardiac stress and how this response differs in those with vascular disease. We measured a comprehensive panel of circulating cell-derived microparticles by a standardized flow cytometric protocol in 119 patients referred for stress echocardiography. Procoagulant, platelet, erythrocyte, and endothelial but not leukocyte, granulocyte, or monocyte-derived microparticles were elevated immediately after a standardized dobutamine stress echocardiogram and decreased after 1 hour. Twenty-five patients developed stress-induced wall motion abnormalities suggestive of myocardial ischemia. They had similar baseline microparticle levels to those who did not develop ischemia, but, interestingly, their microparticle levels did not change during stress. Furthermore, no stress-induced increase was observed in those without inducible ischemia but with a history of vascular disease. Fourteen patients subsequently underwent coronary angiography. A microparticle rise during stress echocardiography had occurred only in those with normal coronary arteries. Procoagulant, platelet, erythrocyte, and endothelial microparticles are released during cardiac stress and then clear from the circulation during the next hour. This stress-induced rise seems to be a normal physiological response that is diminished in those with vascular disease.
Product Description
Dobutamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dobutamine hydrochloride, ≥98%