MilliporeSigma
  • Prognostic model predicting metastatic castration-resistant prostate cancer survival in men treated with second-line chemotherapy.
Prognostic model predicting metastatic castration-resistant prostate cancer survival in men treated with second-line chemotherapy.
Journal of the National Cancer Institute (2013-10-19)
Susan Halabi, Chen-Yen Lin, Eric J Small, Andrew J Armstrong, Ellen B Kaplan, Daniel Petrylak, Cora N Sternberg, Liji Shen, Stephane Oudard, Johann de Bono, Oliver Sartor
ABSTRACT
Several prognostic models for overall survival (OS) have been developed and validated in men with metastatic castration-resistant prostate cancer (mCRPC) who receive first-line chemotherapy. We sought to develop and validate a prognostic model to predict OS in men who had progressed after first-line chemotherapy and were selected to receive second-line chemotherapy. Data from a phase III trial in men with mCRPC who had developed progressive disease after first-line chemotherapy (TROPIC trial) were used. The TROPIC was randomly split into training (n = 507) and testing (n = 248) sets. Another dataset consisting of 488 men previously treated with docetaxel (SPARC trial) was used for external validation. Adaptive least absolute shrinkage and selection operator selected nine prognostic factors of OS. A prognostic score was computed from the regression coefficients. The model was assessed on the testing and validation sets for its predictive accuracy using the time-dependent area under the curve (tAUC). The nine prognostic variables in the final model were Eastern Cooperative Oncology Group performance status, time since last docetaxel use, measurable disease, presence of visceral disease, pain, duration of hormonal use, hemoglobin, prostate specific antigen, and alkaline phosphatase. The tAUCs for this model were 0.73 (95% confidence interval [CI] = 0.72 to 0.74) and 0.70 (95% CI = 0.68 to 0.72) for the testing and validation sets, respectively. A prognostic model of OS in the postdocetaxel, second-line chemotherapy, mCRPC setting was developed and externally validated. This model incorporates novel prognostic factors and can be used to provide predicted probabilities for individual patients and to select patients to participate in clinical trials on the basis of their prognosis. Prospective validation is needed.
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Mitoxantrone dihydrochloride, ≥97% (HPLC)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host