MilliporeSigma
Diagnosis and treatment of Hodgkin's lymphoma: at times a challenge.
Journal of the College of Physicians and Surgeons--Pakistan : JCPSP (2013-10-12)
Nadeem Zia Abbasi, Mir Jalal-ud-Din, Zainab Zahur, Amjad Aziz Khan, Abdul Samad Sheikh, Furqan Ali, Khalid Hussain Memon
ABSTRACT
Hodgkin's lymphoma has been traditionally defined as a hematopoietic neoplasm composed of diagnostic Reed-Sternberg cells. More than 70% of the cases involve cervical or supraclavicular lymph nodes. Isolated sub-diaphragmatic lymphadenopathy or organ involvement is rare. We present the case of Hodgkin's lymphoma in a 51 years old female, who presented with obstructive jaundice and lymphadenopathy, empirically treated previously as a case of tuberculosis. Chemotherapy with modified ABVD protocol was given with dose modification according to LFT's. Her liver functions returned to normal levels after the first cycle. The main purpose of reporting the case is to stress definitive diagnosis of the disease before initiating treatment and the modified chemotherapy regimen used in this infrequent presentation of the disease.
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture