MilliporeSigma
  • Simvastatin for cognitive deficits and behavioural problems in patients with neurofibromatosis type 1 (NF1-SIMCODA): a randomised, placebo-controlled trial.
Simvastatin for cognitive deficits and behavioural problems in patients with neurofibromatosis type 1 (NF1-SIMCODA): a randomised, placebo-controlled trial.
The Lancet. Neurology (2013-10-05)
Thijs van der Vaart, Ellen Plasschaert, André B Rietman, Marleen Renard, Rianne Oostenbrink, Annick Vogels, Marie-Claire Y de Wit, Mie-Jef Descheemaeker, Yvonne Vergouwe, Coriene E Catsman-Berrevoets, Eric Legius, Ype Elgersma, Henriëtte A Moll
ABSTRACT
Neurofibromatosis type 1 is a common genetic disorder characterised by neurocutaneous manifestations and cognitive and behavioural problems. Statins were shown to reduce analogous learning deficits in a mouse model of the disease, but a short-term trial in humans was inconclusive. We aimed to assess the use of simvastatin for the improvement of cognitive and behavioural deficits in children with neurofibromatosis type 1 for 12 months. In this randomised, double-masked, placebo-controlled trial, we recruited children with genetically confirmed neurofibromatosis type 1 aged 8-16 years from two national referral centres in the Netherlands and Belgium. Those with symptomatic CNS abnormalities or on neurotropic medication, including stimulants, were excluded. Eligible patients were randomly assigned (1:1) via a computer-generated, permuted-block list to simvastatin (10 mg per day in month 1, 20 mg per day in month 2, and 20-40 mg per day in months 3-12) or placebo for 12 months. Investigators, participants, and parents were masked to treatment assignment. Primary outcome measures were full-scale intelligence (Wechsler intelligence scale for children), attention problems (child behaviour checklist, parent-rated [CBCL]), and internalising behavioural problems (CBCL). We did intention-to-treat analyses (of all patients who had outcome data) using linear regression of the 12 month outcome scores, adjusted for baseline performance. This trial is registered with the Netherlands Trial Register, number NTR2150. We randomly assigned 84 children to a treatment group (43 to simvastatin, 41 to placebo) between March 9, 2010, and March 6, 2012. We did not assess outcomes in two patients in the placebo group because they needed additional drug therapy. Simvastatin for 12 months had no effect on full-scale intelligence (treatment effect compared with placebo -1·3 IQ points [95% CI -3·8 to 1·3]; p=0·33), attention problems (-1·6 T-score points [-4·3 to 1·0]; p=0·23), and internalising behavioural problems (-0·1 T-score points [-3·3 to 3·1]; p=0·96). 38 (88%) of 43 patients on simvastatin and 39 (95%) of 41 patients on placebo reported adverse events, which were serious in two and four patients, respectively. 12 month simvastatin treatment did not ameliorate cognitive deficits or behavioural problems in children with neurofibromatosis type 1. The use of 20-40 mg simvastatin per day for cognitive enhancement in children with neurofibromatosis type 1 is not recommended. The Netherlands Organization for Health Research and Development (ZonMw), Research Foundation Flanders (FWO-Vlaanderen), Marguerite-Marie Delacroix Foundation, and the Dutch Neurofibromatosis Association (NFVN).
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Simvastatin, European Pharmacopoeia (EP) Reference Standard