MilliporeSigma
  • Tracking the establishment of local endemic populations of an emergent enteric pathogen.
Tracking the establishment of local endemic populations of an emergent enteric pathogen.
Proceedings of the National Academy of Sciences of the United States of America (2013-10-02)
Kathryn E Holt, Tran Vu Thieu Nga, Duy Pham Thanh, Ha Vinh, Dong Wook Kim, My Phan Vu Tra, James I Campbell, Nguyen Van Minh Hoang, Nguyen Thanh Vinh, Pham Van Minh, Cao Thu Thuy, Tran Thi Thu Nga, Corinne Thompson, Tran Thi Ngoc Dung, Nguyen Thi Khanh Nhu, Phat Voong Vinh, Pham Thi Ngoc Tuyet, Hoang Le Phuc, Nguyen Thi Nam Lien, Bui Duc Phu, Nguyen Thi Thuy Ai, Nguyen Manh Tien, Nguyen Dong, Christopher M Parry, Tran Tinh Hien, Jeremy J Farrar, Julian Parkhill, Gordon Dougan, Nicholas R Thomson, Stephen Baker
ABSTRACT
Shigella sonnei is a human-adapted pathogen that is emerging globally as the dominant agent of bacterial dysentery. To investigate local establishment, we sequenced the genomes of 263 Vietnamese S. sonnei isolated over 15 y. Our data show that S. sonnei was introduced into Vietnam in the 1980s and has undergone localized clonal expansion, punctuated by genomic fixation events through periodic selective sweeps. We uncover geographical spread, spatially restricted frontier populations, and convergent evolution through local gene pool sampling. This work provides a unique, high-resolution insight into the microevolution of a pioneering human pathogen during its establishment in a new host population.
Product Description
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Ciprofloxacin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material