MilliporeSigma
  • Probing structural requirements of positive allosteric modulators of the M4 muscarinic receptor.
Probing structural requirements of positive allosteric modulators of the M4 muscarinic receptor.
Journal of medicinal chemistry (2013-10-01)
Tracey Huynh, Celine Valant, Ian T Crosby, Patrick M Sexton, Arthur Christopoulos, Ben Capuano
ABSTRACT
The M4 mAChR is implicated in several CNS disorders and possesses an allosteric binding site for which ligands modulating the affinity and/or efficacy of ACh may be exploited for selective receptor targeting. We report the synthesis of a focused library of putative M4 PAMs derived from VU0152100 and VU10005. These compounds investigate the pharmacological effects of previously identified methoxy and fluoro substituents, providing useful estimates of affinity (KB), cooperativity (αβ), and direct agonist properties (τB).
Sigma-Aldrich
Acetylcholine chloride, pkg of 150 mg (per vial)
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC), free-flowing, Redi-Dri
Sigma-Aldrich
Acetylcholine iodide, ≥97%
Sigma-Aldrich
Acetylcholine bromide, ≥99%