MilliporeSigma
  • Multimodal surgical guidance towards the sentinel node in vulvar cancer.
Multimodal surgical guidance towards the sentinel node in vulvar cancer.
Gynecologic oncology (2013-09-21)
H M Mathéron, N S van den Berg, O R Brouwer, G H Kleinjan, W J van Driel, J W Trum, E Vegt, G Kenter, F W B van Leeuwen, R A Valdés Olmos
ABSTRACT
Conventional sentinel node (SN) mapping is performed by injecting a radiocolloid followed by lymphoscintigraphy (and SPECT/CT imaging). An extra intraoperative injection with blue dye can then allow for optical identification of the SN. In order to improve the current clinical standard, the hybrid tracer indocyanine green (ICG)-(99m)Tc-nanocolloid was introduced, a tracer that is both radioactive and fluorescent. This feasibility study aimed to evaluate the value of a multimodal-based SN biopsy in vulvar cancer. Fifteen patients with vulvar cancer (29 groins) scheduled for SN biopsy were peritumorally injected with ICG-(99m)Tc-nanocolloid followed by lymphoscintigraphy and SPECT/CT imaging to identify the SNs. In thirteen patients, shortly before the start of the operation, blue dye was intradermally injected around the lesion. SNs were harvested using a combination of radiotracing, fluorescence imaging, and optical blue dye detection. A portable gamma camera was used before and after SN excision to confirm excision of the preoperatively defined SNs. Preoperative lymphoscintigraphy and SPECT/CT imaging visualized drainage to 39 SNs in 28 groins. During the operation, 98% (ex vivo 100%) of the SNs were radioactive. With fluorescence imaging 96% of the SNs (ex vivo 100%) could be visualized. Only 65% of the SNs had stained blue at the time of excision. ICG-(99m)Tc-nanocolloid can be used for preoperative SN identification and enables multimodal (radioactive and fluorescent) surgical guidance in patients with vulvar cancer. The addition of fluorescence-based optical guidance offers more effective SN visualization compared to blue dye.
Sigma-Aldrich
Cardiogreen, for microscopy
Sigma-Aldrich
Cardiogreen, polymethine dye