MilliporeSigma
  • The receptor antagonist picotamide inhibits adrenergic and thromboxane-induced contraction of hyperplastic human prostate smooth muscle.
The receptor antagonist picotamide inhibits adrenergic and thromboxane-induced contraction of hyperplastic human prostate smooth muscle.
American journal of physiology. Renal physiology (2013-09-21)
Martin Hennenberg, Marijan Miljak, Daniel Herrmann, Frank Strittmatter, Sebastian Walther, Beata Rutz, Yasemin Hocaoglu, Thomas Kunit, Andrea Schreiber, Karl-Erik Andersson, Christian G Stief, Christian Gratzke
ABSTRACT
Inhibition of prostate smooth muscle contraction is an important strategy for medical treatment of lower urinary tract symptoms (LUTS). Besides α1-adrenoceptors, prostate smooth muscle contraction is induced by activation of thromboxane (TXA2) receptors (TXA2-R). Here, we examined the effects of the TXA2-R antagonist picotamide on contraction of human prostate tissue. Prostate tissues were obtained from radical prostatectomy. The effects of picotamide (300 μM), L-665,240 (3 μM), and seratrodast (3 μM) on U46619-, electric field stimulation- (EFS-), phenylephrine-, and norepinephrine-induced contractions were studied in organ baths. Expression of TXA2-R and TXA2 synthase (TXS) was examined by fluorescence stainings. Picotamide, seratrodast, and L-655,240 inhibited concentration-dependent contractions induced by the TXA2 analog U46619. Picotamide, but not seratrodast or L-655,240, inhibited frequency-dependent contractions induced by EFS. Picotamide inhibited concentration-dependent contractions induced by norepinephrine or by the selective α1-adrenoceptor agonist phenylephrine. In prostate strips, where only submaximal contraction by a low dose of phenylephrine was induced, application of U46619 raised tone to maximum phenylephrine-induced tension. Immunoreactivity for TXA2-R and TXS was observed in the stroma and in epithelial cells of glands. Colocalization of both immunoreactivites was observed with the smooth muscle markers calponin and α-smooth muscle actin, with the epithelial marker pan-cytokeratin, and with prostate-specific antigen in the stroma and glands. The receptor antagonist picotamide inhibits α1-adrenergic, TXA2-mediated, and EFS-induced contractions in the human prostate. To the best of our knowledge, this is the first antagonist able to inhibit two different contraction systems in the prostate.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Cy3 conjugate, Chemicon®, from goat
Sigma-Aldrich
Picotamide