MilliporeSigma
  • Cognitive-behavioral therapy vs risperidone for augmenting serotonin reuptake inhibitors in obsessive-compulsive disorder: a randomized clinical trial.
Cognitive-behavioral therapy vs risperidone for augmenting serotonin reuptake inhibitors in obsessive-compulsive disorder: a randomized clinical trial.
JAMA psychiatry (2013-09-13)
Helen Blair Simpson, Edna B Foa, Michael R Liebowitz, Jonathan D Huppert, Shawn Cahill, Michael J Maher, Carmen P McLean, James Bender, Sue M Marcus, Monnica T Williams, Jamie Weaver, Donna Vermes, Page E Van Meter, Carolyn I Rodriguez, Mark Powers, Anthony Pinto, Patricia Imms, Chang-Gyu Hahn, Raphael Campeas
ABSTRACT
Obsessive-compulsive disorder (OCD) is one of the world's most disabling illnesses according to the World Health Organization. Serotonin reuptake inhibitors (SRIs) are the only medications approved by the Food and Drug Administration to treat OCD, but few patients achieve minimal symptoms from an SRI alone. In such cases, practice guidelines recommend adding antipsychotics or cognitive-behavioral therapy consisting of exposure and ritual prevention (EX/RP). To compare the effects of these 2 SRI augmentation strategies vs pill placebo for the first time, to our knowledge, in adults with OCD. A randomized clinical trial (conducted January 2007-August 2012) at 2 academic outpatient research clinics that specialize in OCD and anxiety disorders. Patients (aged 18-70 years) were eligible if they had OCD of at least moderate severity despite a therapeutic SRI dose for at least 12 weeks prior to entry. Of 163 who were eligible, 100 were randomized (risperidone, n = 40; EX/RP, n = 40; and placebo, n = 20), and 86 completed the trial. While continuing their SRI at the same dose, patients were randomized to the addition of 8 weeks of risperidone (up to 4 mg/d), EX/RP (17 sessions delivered twice weekly), or pill placebo. Independent assessments were conducted every 4 weeks. The Yale-Brown Obsessive Compulsive Scale (Y-BOCS) to measure OCD severity. Patients randomized to EX/RP had significantly greater reduction in week 8 Y-BOCS scores based on mixed-effects models (vs risperidone: mean [SE], -9.72 [1.38]; P < .001 vs placebo: mean [SE], -10.10 [1.68]; P < .001). Patients receiving risperidone did not significantly differ from those receiving placebo (mean [SE], -0.38 [1.72]; P = .83). More patients receiving EX/RP responded (Y-BOCS score decrease ≥25%: 80% for EX/RP, 23% for risperidone, and 15% for placebo; P < .001). More patients receiving EX/RP achieved minimal symptoms (Y-BOCS score ≤12: 43% for EX/RP, 13% for risperidone, and 5% for placebo; P = .001). Adding EX/RP was also superior to risperidone and placebo in improving insight, functioning, and quality of life. Adding EX/RP to SRIs was superior to both risperidone and pill placebo. Patients with OCD receiving SRIs who continue to have clinically significant symptoms should be offered EX/RP before antipsychotics given its superior efficacy and less negative adverse effect profile. clinicaltrials.gov Identifier: NCT00389493.
Product Description
Risperidone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Risperidone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder
Risperidone, European Pharmacopoeia (EP) Reference Standard