MilliporeSigma
  • Synthesis of fused-β-lactams through selective gold-catalyzed oxycyclization of dioxolane-tethered enynes.
Synthesis of fused-β-lactams through selective gold-catalyzed oxycyclization of dioxolane-tethered enynes.
The Journal of organic chemistry (2013-09-07)
Benito Alcaide, Pedro Almendros, Teresa Martínez del Campo, M Rosario Torres
ABSTRACT
The gold-catalyzed preparation of 2-azetidinone-fused oxacycles was accomplished from β-lactam-linked enynes through heterocyclization reaction taking advantage of the acetonide pendant group. While the synthesis of fused tetrahydrofuran-β-lactams from 1,3-enynes could be considered as an unusual metal-catalyzed cyclization of enynols, α-alkoxy dioxolane-tethered 1,3-enynes exclusively undergo bis-oxycyclization to afford tricyclic bridged acetals.
Sigma-Aldrich
1,3-Dioxolane, anhydrous, contains ~75 ppm BHT as inhibitor, 99.8%
Sigma-Aldrich
1,3-Dioxolane, ReagentPlus®, contains ~75 ppm BHT as inhibitor, 99%