MilliporeSigma
  • Behavior of human immunoglobulin G adsorption onto immobilized Cu(II) affinity hollow-fiber membranes.
Behavior of human immunoglobulin G adsorption onto immobilized Cu(II) affinity hollow-fiber membranes.
Journal of molecular recognition : JMR (2013-09-03)
Mariana Borsoi-Ribeiro, Igor Tadeu Lazzarotto Bresolin, Mookambeswaran Vijayalakshmi, Sônia Maria Alves Bueno
ABSTRACT
Iminodiacetic acid (IDA) and tris(2-aminoethyl)amine (TREN) chelating ligands were immobilized on poly(ethylene vinyl alcohol) (PEVA) hollow-fiber membranes after activation with epichlorohydrin or butanediol diglycidyl ether (bisoxirane). The affinity membranes complexed with Cu(II) were evaluated for adsorption of human immunoglobulin G (IgG). The effects of matrix activation and buffer system on adsorption of IgG were studied. Isotherms of batch IgG adsorption onto finely cut membranes showed that neither of the chelates, IDA-Cu(II) or TREN-Cu(II), had a Langmuirean behavior with negative cooperativity for IgG binding. A comparison of equilibrium and dynamic maximum capacities showed that the dynamic capacity for a mini-cartridge in a cross-flow filtration mode (52.5 and 298.4 mg g(-1) dry weight for PEVA-TREN-Cu(II) and PEVA-IDA-Cu(II), respectively) was somewhat higher than the equilibrium capacity (9.2 and 73.3 mg g(-1) dry weight for PEVA-TREN-Cu(II) and PEVA-IDA-Cu(II), respectively). When mini-cartridges were used, the dynamic adsorption capacity of IDA-Cu(II) was the same for both mini-cartridge and agarose gel.
Product Description
Supelco
(±)-Epichlorohydrin, analytical standard
Sigma-Aldrich
Tris(2-aminoethyl)amine, 96%
Sigma-Aldrich
(±)-Epichlorohydrin, purum, ≥99% (GC)
Sigma-Aldrich
Iminodiacetic acid, purum, ≥98.0% (T)
Sigma-Aldrich
(S)-(+)-Epichlorohydrin, 98%
Sigma-Aldrich
Iminodiacetic acid, 98%
Sigma-Aldrich
Poly(vinyl alcohol-co-ethylene), ethylene 32 mol %