MilliporeSigma
  • Neuropathic pain following sagittal split ramus osteotomy of the mandible: prevalence, risk factors, and clinical course.
Neuropathic pain following sagittal split ramus osteotomy of the mandible: prevalence, risk factors, and clinical course.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2013-09-03)
Érica C Marchiori, Jacob S Barber, W Bradford Williams, Peter Q Bui, Felice S O'Ryan
ABSTRACT
To estimate the prevalence of, risk factors for, and clinical course of neuropathic pain (NPP) after sagittal split ramus osteotomy (SSRO) of the mandible in a large cohort of patients. A retrospective cohort of all patients who underwent SSRO at 2 medical centers within Kaiser Permanente Northern California from January 2007 through September 2012 was assembled. Demographic, clinical, and surgical factors were abstracted from medical records and relevant comorbidities were identified. The prevalence of NPP in the cohort was calculated and the clinical signs, symptoms, temporal characteristics, and treatment response in affected patients were noted. The authors identified 1,778 patients who underwent SSRO and excluded 107 patients according to predefined criteria. The remaining 1,671 patients had a median age of 24 years (interquartile range, 19 to 35 yr) and 62.4% were women. Seven patients developed NPP after SSRO, which was an overall prevalence of 0.42%. All patients with NPP in this cohort were women and had a median age of 49 years. The risk factors for developing NPP after this surgery were older age (P = .0098), depression (P = .0100), and female gender. NPP developed an average of 30 days postoperatively (range, 18 to 56 days) and persisted for a median duration of 52 days (range, 30 to 69.5 days). All patients responded favorably to anticonvulsant (n = 6) or tricyclic (n = 1) medications, and no patients developed chronic postsurgical pain. NPP was an infrequent complication after SSRO, occurring in 1 of 238 patients in this cohort. The short duration and positive response to medication are reassuring findings. The results of this investigation highlight the need for prospective studies to further understand the spectrum of postoperative NPP.
Product Description
Supelco
Gabapentin solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Supelco
Gabapentin, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Gabapentin, solid
Pricing and availability is not currently available.