MilliporeSigma
  • Construction of multifunctional modules for drug discovery: synthesis of novel thia/oxa-azaspiro[3.4]octanes.
Construction of multifunctional modules for drug discovery: synthesis of novel thia/oxa-azaspiro[3.4]octanes.
Organic letters (2013-09-03)
Dong Bo Li, Mark Rogers-Evans, Erick M Carreira
ABSTRACT
New classes of thia/oxa-azaspiro[3.4]octanes are synthesized through the implementation of robust and step-economic routes. The targeted spirocycles have been designed to act as novel, multifunctional, and structurally diverse modules for drug discovery. Furthermore, enantioselective approaches to the spirocycles are reported.
Sigma-Aldrich
Oxytocin
Sigma-Aldrich
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)
Oxytocin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oxytocin acetate salt hydrate, ≥97% (HPLC)