MilliporeSigma
  • Kaposi's sarcoma-associated herpesvirus-positive primary effusion lymphoma tumor formation in NOD/SCID mice is inhibited by neomycin and neamine blocking angiogenin's nuclear translocation.
Kaposi's sarcoma-associated herpesvirus-positive primary effusion lymphoma tumor formation in NOD/SCID mice is inhibited by neomycin and neamine blocking angiogenin's nuclear translocation.
Journal of virology (2013-08-30)
Virginie Bottero, Sathish Sadagopan, Karen E Johnson, Sujoy Dutta, Mohanan Valiya Veettil, Bala Chandran
ABSTRACT
Angiogenin (ANG) is a 14-kDa multifunctional proangiogenic secreted protein whose expression level correlates with the aggressiveness of several tumors. We observed increased ANG expression and secretion in endothelial cells during de novo infection with Kaposi's sarcoma-associated herpesvirus (KSHV), in cells expressing only latency-associated nuclear antigen 1 (LANA-1) protein, and in KSHV latently infected primary effusion lymphoma (PEL) BCBL-1 and BC-3 cells. Inhibition of phospholipase Cγ (PLCγ) mediated ANG's nuclear translocation by neomycin, an aminoglycoside antibiotic (not G418-neomicin), resulted in reduced KSHV latent gene expression, increased lytic gene expression, and increased cell death of KSHV(+) PEL and endothelial cells. ANG detection in significant levels in KS and PEL lesions highlights its importance in KSHV pathogenesis. To assess the in vivo antitumor activity of neomycin and neamine (a nontoxic derivative of neomycin), BCBL-1 cells were injected intraperitoneally into NOD/SCID mice. We observed significant extended survival of mice treated with neomycin or neamine. Markers of lymphoma establishment, such as increases in animal body weight, spleen size, tumor cell spleen infiltration, and ascites volume, were observed in nontreated animals and were significantly diminished by neomycin or neamine treatments. A significant decrease in LANA-1 expression, an increase in lytic gene expression, and an increase in cleaved caspase-3 were also observed in neomycin- or neamine-treated animal ascitic cells. These studies demonstrated that ANG played an essential role in KSHV latency maintenance and BCBL-1 cell survival in vivo, and targeting ANG function by neomycin/neamine to induce the apoptosis of cells latently infected with KSHV is an attractive therapeutic strategy against KSHV-associated malignancies.
Product Description
Supelco
Neomycin trisulfate salt hydrate, VETRANAL®, analytical standard
Neomycin sulphate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Neomycin trisulfate salt hydrate, suitable for plant cell culture
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-AS, 50-100 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type X-A, ≥90% (SDS-PAGE), ≥70 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Ribonuclease B from bovine pancreas, BioReagent, ≥50 Kunitz units/mg protein, ≥80% (SDS-PAGE)
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Neomycin trisulfate salt hydrate, meets USP testing specifications, powder
Sigma-Aldrich
Ribonuclease A from bovine pancreas, 90%, powder, white, ~80 U/mg
Sigma-Aldrich
Ribonuclease A from bovine pancreas, 4×cryst., ~70 U/mg (acc. to Kunitz)
Neamine, European Pharmacopoeia (EP) Reference Standard